Skip to Content
Merck
All Photos(1)

Key Documents

T56308

Sigma-Aldrich

2,6-Dichlorobenzyl chloride

97%

Synonym(s):

α,2,6-Trichlorotoluene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$46.30

$46.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$46.30

About This Item

Linear Formula:
Cl2C6H3CH2Cl
CAS Number:
Molecular Weight:
195.47
Beilstein:
472282
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$46.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

bp

117-119 °C/14 mmHg (lit.)

mp

36-39 °C (lit.)

SMILES string

ClCc1c(Cl)cccc1Cl

InChI

1S/C7H5Cl3/c8-4-5-6(9)2-1-3-7(5)10/h1-3H,4H2

InChI key

LBOBESSDSGODDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Chu et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 19(2), 183-191 (1984-03-01)
Groups of 10 male and 10 female rats were fed alpha,alpha,alpha-, alpha,2,6- or 2,3,6- trichlorotoluene (TCT) in their diet at 0, 0.5, 5.0, 50 or 500 ppm for 28 days. Growth rate and food consumption were not affected by treatment.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service