Skip to Content
Merck
All Photos(2)

Key Documents

105740

Sigma-Aldrich

α,α′-Dichloro-p-xylene

98%

Synonym(s):

1,4-Bis(chloromethyl)benzene, p-Xylylene dichloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$47.70
100 G
$138.00

$47.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$47.70
100 G
$138.00

About This Item

Linear Formula:
C6H4(CH2Cl)2
CAS Number:
Molecular Weight:
175.06
Beilstein:
774706
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$47.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

bp

254 °C (lit.)

mp

98-101 °C (lit.)

functional group

chloro

SMILES string

ClCc1ccc(CCl)cc1

InChI

1S/C8H8Cl2/c9-5-7-1-2-8(6-10)4-3-7/h1-4H,5-6H2

InChI key

ZZHIDJWUJRKHGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α,α′-Dichloro-p-xylene undergoes phase transfer catalysed polycondensation with t-butylcyanoacetate to form carbon-carbon chain polymer[1].

Application

α,α′-Dichloro-p-xylene is a monomer used in preparation of poly(p-phenylene vinylene(PPV) nanotubes and nanorods by chemical vapor deposition (CVD) polymerization method[2]. α,α′-Dichloro-p-xylene was used in a study to investigate the effect of CVD monomer selection and reaction conditions on the resulting PPV polymer layer composition and luminescence properties[3].

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of carbon-carbon chain polymer by phase-transfer-catalyzed polycondensation of α, α'-dichloro-p-xylene with t-butyl cyanoacetate.
Imai Y, et al.
Journal of Polymer Science - Part C: Polymer Letters, 19(4), 205-210 (1981)
Preparation of PPV nanotubes and nanorods and carbonized products derived therefrom.
Kim K and Jin J.
Nano Letters, 1(11), 205-210 (1981)
Poly (p-phenylene vinylene) prepared by chemical vapor deposition: Influence of monomer selection and reaction conditions on film composition and luminescence properties.
Vaeth KM and Jensen KF.
Macromolecules, 31(20), 6789-6793 (1998)
Contact dermatitis caused by "chloromethyl heterocyclic intermediates".
H Ippen
Contact dermatitis, 16(1), 52-52 (1987-01-01)
Jing-Wei Xu et al.
Colloids and surfaces. B, Biointerfaces, 133, 148-155 (2015-06-22)
Electrospun nanofibers with antibacterial activity are greatly promising for medical treatment and water purification. Herein we report antibacterial nanofibers electrospun from a series of poly(dimethylamino ethyl methacrylate-co-alkyl methacrylates) (poly(DMAEMA-co-AMA)) and to distinguish the effects of free and cross-linked cations derived

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service