Key Documents
SYNS735060
Synthware sublimator, complete
60 mL, complete
Synonym(s):
Synthware S735060, sublimation apparatus
Select a Size
1 EA
$1,080.00
$1,080.00
Estimated to ship onMay 29, 2025
Select a Size
Change View
1 EA
$1,080.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
$1,080.00
Estimated to ship onMay 29, 2025
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S735060
capacity
60 mL
Related Categories
General description
A grease-free, two-piece sublimation apparatus consisting of an upper water cooled condenser and lower vessel to hold the material to be sublimed. The sublimate collects on the lower end of the condenser which has a flat bottom. Condenser is constructed using a grease-free, threaded valve. Connection of the condenser and lower vessel via an O-ring joint enables the two parts to be gently separated for maximum product recovery. Complete apparatus consists of the cold finger condenser, lower vessel, Viton® O-ring and stainless steel pinch clamp.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)