Key Documents
SYNS580012
Synthware 3-way high vacuum valve
size 0-12 mm
Synonym(s):
Synthware S580012
Select a Size
1 EA
$308.00
$308.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$308.00
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
$308.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S580012
General description
With two 9 mm O.D. sidearms sealed to the barrel at a 90° angle to one another. The non-rotating PTFE plug has three O-rings, the tip O-ring making the primary seal on the precision formed tapered seat.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)