Key Documents
SYNS510217
Synthware T-bore glass stopcock Entire assembly
bore size 2 mm, plug size 12/30
Synonym(s):
Synthware S510217, T-shape stopcock
Select a Size
1 EA
$23.10
$23.10
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$23.10
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$23.10
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S510217
bore size
2 mm
Related Categories
General description
With solid glass plug. With constant pressure retention spring and clip.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)