Key Documents
SYNM522403
Synthware vacuum manifold with high vacuum valves
number of ports, 3, Hose Connection: left
Synonym(s):
Synthware M522403, vacuum manifold, gas manifold
Select a Size
1 EA
$616.00
$616.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$616.00
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
$616.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
feature
number of ports 3
packaging
pkg of 1 ea
manufacturer/tradename
Synthware M522403
General description
With a serrated hose connection(10 mm O.D.) at left hand side. Main tube is 22 mm O.D. and the distance between valves is 100 mm center to center.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)