Key Documents
SYNF419100
Synthware two neck round bottom flask with angled side neck
100 mL, center joint: ST/NS 24/40, side joint: ST/NS 19/22
Synonym(s):
Synthware F419100, flask, two-neck flask, round bottom flask
Select a Size
1 EA
$159.00
$159.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$159.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
$159.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass flask
round bottom
two-neck (angled side neck)
Joint
center joint: ST/NS24/40
side joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F419100
capacity
100 mL
100 mL
General description
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)