Documents
SYNA392440
Synthware transfer valve adapter
joint: ST/NS 24/40
Synonym(s):
Synthware A392440, transfer adapter with valve
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A392440
Related Categories
General description
The 0-4 mm high vacuum valves have two Viton® O-rings on the shaft and an integral PTFE O-ring on the tip.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company