Key Documents
SYND351403
Synthware cow type graduated distillation receiver
joint: ST/NS 14/20
Synonym(s):
Synthware D351403, cow type distilling receiver
Select a Size
1 EA
$184.00
$184.00
Estimated to ship onApril 29, 2025
Select a Size
Change View
1 EA
$184.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
$184.00
Estimated to ship onApril 29, 2025
Joint
joint: ST/NS14/20
manufacturer/tradename
D351403
Related Categories
General description
Synthware distillation cow receiver features a standard taper joint (14/20) and four tubes graduated from 0-3mL in 0.2mL subdivisions.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)