Skip to Content
Merck
All Photos(1)

Documents

75061

Sigma-Aldrich

Tri-tert-butyl 1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetate

≥95.0% (CHN)

Synonym(s):

DOTA-tri-t-Bu-ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H52N4O8
CAS Number:
Molecular Weight:
572.73
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (CHN)

form

lumps

SMILES string

CC(C)(C)OC(=O)CN1CCN(CCN(CCN(CC1)CC(=O)OC(C)(C)C)CC(=O)OC(C)(C)C)CC(O)=O

InChI

1S/C28H52N4O8/c1-26(2,3)38-23(35)19-30-12-10-29(18-22(33)34)11-13-31(20-24(36)39-27(4,5)6)15-17-32(16-14-30)21-25(37)40-28(7,8)9/h10-21H2,1-9H3,(H,33,34)

InChI key

RVUXZXMKYMSWOM-UHFFFAOYSA-N

Related Categories

Application

Tri-tert-butyl 1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetate (tri BOC-DOTA) can be employed as a reactant to prepare:
  • Gadolinium ion functionalized with DOTA-LAE (lactobionic acid-ethylenediamine), which is used as a contrast agent for magnetic resonance imaging (MRI).
  • DOTA-conjugates of ursolic acid and ZD2 peptide 64Cu-DOTA conjugates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin Poret et al.
Biomolecules, 10(3) (2020-03-25)
Overexpression of G protein-coupled receptors (GPCRs) in tumours is widely used to develop GPCR-targeting radioligands for solid tumour imaging in the context of diagnosis and even treatment. The human vasoactive neuropeptide urotensin II (hUII), which shares structural analogies with somatostatin
Preparation and evaluation of ZD2 peptide 64Cu-DOTA conjugate as a positron emission tomography probe for detection and characterization of prostate cancer
Han Z, et al.
ACS Omega, 4(1), 1185-1190 (2019)
Synthesis of a gadolinium based-macrocyclic MRI contrast agent for effective cancer diagnosis
Jeong Y and Na K
Biomaterials research, 22(1), 17-17 (2018)
Synthesis and cytotoxicity evaluation of DOTA-conjugates of ursolic acid
Kahnt, M, et al.
Molecules (Basel), 24(12), 2254-2254 (2019)
Lubinda Mbundi et al.
Nanomaterials (Basel, Switzerland), 8(6) (2018-06-06)
The control of osteoblast/osteoclast cross-talk is crucial in the bone remodelling process and provides a target mechanism in the development of drugs for bone metabolic diseases. Osteoprotegerin is a key molecule in this biosignalling pathway as it inhibits osteoclastogenesis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service