Key Documents
SYND191922
Synthware distilling receiver with hose connection
joint: ST/NS 19/22
Synonym(s):
Synthware D191922, glass distilling receiver
Select a Size
1 EA
$159.00
$159.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$159.00
About This Item
UNSPSC Code:
41104812
NACRES:
NB.51
$159.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware D191922
Related Categories
General description
Cow-type design having three receivers spaced 40o apart. 105o angle between top outer joint and lower inner joints.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)