Key Documents
SYNC101922
Synthware Vigreux distillation column
joint: ST/NS 19/22, L 120 mm
Synonym(s):
Synthware C101922, distilling column, fractional column
Select a Size
1 EA
$85.00
$85.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$85.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$85.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C101922
L
120 mm
Related Categories
General description
Column length is measured from the highest to lowest indent in the column.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)