Key Documents
SYNA521420
Synthware inlet or thermometer adapter with compression cap
joint: ST/NS 14/20, O.D. 5.0-7.5 mm
Synonym(s):
Synthware A521420, thermometer inlet adapter
Select a Size
1 EA
$67.70
$67.70
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$67.70
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$67.70
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A521420
O.D.
5.0-7.5 mm
General description
With a compression cap and Viton® O-ring to form a vacuum tight seal. To use it as a syringe port, simply replace the O-ring with PTFE -lined septum. Cap accommodates 5.0 - 7.5mm O.D. thermometer. Supplied with one Nylon cap and one Viton® O-ring.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)