Skip to Content
Merck
All Photos(1)

Documents

SLW9830/29M

Pyrex® Filter holder for 25 mm membranes

clip only

Synonym(s):

Pyrex, SciLabware 9830/29M

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123300
NACRES:
NB.43

manufacturer/tradename

9830/29M
SciLabware

Looking for similar products? Visit Product Comparison Guide

General description

  • With porosity grade 3 sintered glass disc to support 25mm diameter membranes
  • Liquids only in contact with chemically resistant borosilicate glass
  • Fully autoclavable

Comprises three components which should be ordered separately
  • Funnel 15ml capacity
  • Support base with sintered glass disc
  • Metal retaining clip

Features and Benefits

• With porosity grade 3 sintered glass disc to support 25 mm diameter membranes• Liquids only in contact with chemically resistant borosilicate glass• Comprises three components which should be ordered separately: - Graduated 15 ml capacity funnel - Support base with sintered glass disc - Metal retaining clip• Fully autoclavable

Legal Information

PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service