Skip to Content
Merck
All Photos(2)

Key Documents

SLW4500/01

Pyrex® Square plastic feet

for 25 mL measuring cylinder

Synonym(s):

Pyrex, SciLabware 4500/01

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$47.60

$47.60


Estimated to ship onMay 01, 2025



Select a Size

Change View
10 EA
$47.60

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

$47.60


Estimated to ship onMay 01, 2025


material

polyethylene

manufacturer/tradename

4500/01
SciLabware

Looking for similar products? Visit Product Comparison Guide

General description

For use with SLW3205 cylinders

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service