Skip to Content
Merck
All Photos(4)

Key Documents

S7556

Sigma-Aldrich

Sulfur trioxide pyridine complex

97%

Synonym(s):

NSC 75831, Pyridine sulfur trioxide complex

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$55.00
100 G
$144.00
1 KG
$796.00

$55.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$55.00
100 G
$144.00
1 KG
$796.00

About This Item

Empirical Formula (Hill Notation):
C5H5NO3S
CAS Number:
Molecular Weight:
159.16
Beilstein:
3704116
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.22

$55.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

reaction suitability

reagent type: oxidant

SMILES string

O=S(=O)=O.c1ccncc1

InChI

1S/C5H5N.O3S/c1-2-4-6-5-3-1;1-4(2)3/h1-5H;

InChI key

UDYFLDICVHJSOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Sulfation agent

Reactant for:
  • Preparation of azido anologs of pregnanolone
  • Sulfate esters of morphine derivatives
  • Gold nanoparticles capped with sulfate-ended ligands as anti-HIV agents
Sulfur trioxide-pyridine (SO3-py) complex can be used to convert the allylic or benzylic hydroxyl group of compounds such as geraniol, farnesol, and 1-indanol into sulfate monoester, which is a good leaving group. This method is highly useful in the total synthesis of sesquiterpenes.[1]
As a sulfating agent, SO3-py complex can be used for the preparation of sulfated-chitins[2], sulfated hyaluronan (HyaS) derivatives[3] and sulfated starch.[4]

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of sulfated-chitins under homogeneous conditions.
Zou Y and Khor E
Carbohydrate Polymers, 77(3), 516-525 (2009)
A method for deoxygenation of allylic and benzylic alcohols.
Corey E J and Achiwa K
The Journal of Organic Chemistry, 34(11), 3667-3668 (1969)
Polysaccharide-based superporous hydrogels with fast swelling and superabsorbent properties.
Kuang J, et al.
Carbohydrate Polymers, 83(1), 284-290 (2011)
Helyn Priscila de Oliveira Barddal et al.
International journal of biological macromolecules, 145, 604-610 (2019-12-31)
Heparin is an extremely important and recognized anticoagulant and antithrombotic agent. Obtained from animal sources and being highly potent, risks of contamination by pathogens and bleeding are some concerns related to heparin use. In the search for alternatives to heparin
Sulfated hyaluronan derivatives reduce the proliferation rate of primary rat calvarial osteoblasts.
Kunze R, et al.
Glycoconjugate Journal, 27(1), 151-158 (2010)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service