Skip to Content
Merck
All Photos(2)

Key Documents

84739

Sigma-Aldrich

Sulfur trioxide triethylamine complex

technical, ≥95% sulfur basis

Synonym(s):

NSC 68185, NSC 8168, Triethylamine sulfur trioxide complex

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$194.00
50 G
$695.00

$194.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$194.00
50 G
$695.00

About This Item

Linear Formula:
(CH3CH2)3N · SO3
CAS Number:
Molecular Weight:
181.25
Beilstein:
3993165
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$194.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical

Quality Level

Assay

≥95% sulfur basis

form

powder

reaction suitability

reagent type: oxidant

mp

~85 °C

functional group

amine

storage temp.

2-8°C

SMILES string

O=S(=O)=O.CCN(CC)CC

InChI

1S/C6H15N.O3S/c1-4-7(5-2)6-3;1-4(2)3/h4-6H2,1-3H3;

InChI key

YYHPEVZFVMVUNJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for reagent for synthesis of:
  • Anticoagulant and antiplatelet persulfated glycosides or flavonoids containing an oligopolysulfated moiety
  • Narciclasine derivatives for use as anticancer agents
  • Sulfate-conjugated methylprednisolone for use as a colon-specific prodrug
  • Tetrasaccharide substrates of heparanase by glycosylation of disaccharides
  • Synthetic heparin oligosaccharide microarrays for analysis of heparin-protein interactions
  • Dexamethasone 21-sulfate sodium for use as a colon-specific prodrug

Other Notes

Reagent used for the sulfation of alcohols and other compounds[1][2]

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deepika Vaidyanathan et al.
Glycobiology, 30(11), 847-858 (2020-04-19)
The chemoenzymatic synthesis of heparin, through a multienzyme process, represents a critical challenge in providing a safe and effective substitute for this animal-sourced anticoagulant drug. D-glucuronyl C5-epimerase (C5-epi) is an enzyme acting on a heparin precursor, N-sulfoheparosan, catalyzing the reversible
Effect of hexadimethrine bromide on the conversion of 4-14C-progesterone into aldosterone and corticosterone by rat adrenals.
A G Fazekas et al.
Steroids, 9(5), 499-506 (1967-05-01)
Cátia Vilas-Boas et al.
Marine drugs, 18(10) (2020-10-01)
Marine biofouling represents a global economic and ecological challenge and few eco-friendly antifouling agents are available. The aim of this work was to establish the proof of concept that a recently synthesized nature-inspired compound (gallic acid persulfate, GAP) can act
Marta Correia-da-Silva et al.
Journal of molecular endocrinology, 61(2), M27-M39 (2018-03-29)
Resveratrol (RSV) is a polyphenolic compound with antioxidant, anti-inflammatory and anti-aging properties partly associated with sirtuin 1 (SIRT1)-activation in the skin. However, poor water solubility may limit RSV efficacy. This work aimed to clarify the interest of a new synthetic
Irshad Ahmad et al.
International journal of biological macromolecules, 119, 360-368 (2018-07-17)
Currently available anticoagulants for prevention and treatment of thrombosis have several limitations, thus, small organic scaffolds that can dissolve clots in vivo in a dose dependent manner with lesser side effects are highly desirable. Here we report the synthesis of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service