Skip to Content
Merck
All Photos(1)

Key Documents

PCL30K

Sigma-Aldrich

PCL Microspheres

30 μm average diameter

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$597.00

$597.00


Estimated to ship onApril 17, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$597.00

About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

$597.00


Estimated to ship onApril 17, 2025


Request a Bulk Order

average diameter

30 μm

storage temp.

-10 to -25°C

Looking for similar products? Visit Product Comparison Guide

General description

PCL particles are made with poly(Ɛ-caprolactone) with a MW of 14,000.To re-constitute:
  • Warm to ambient temperature
  • Add DI water or buffer to achieve desired concentration
  • Vortex and sonicate for 5 minutes to ensure particles are well suspended

10 mg/ml particle suspension in DI water. Sonicate or vortex the particles before each use.

Application

Plain PCL particles for testing system compatibility in drug delivery research. Particles can be used as carriers by attaching drug or biological/chemical entities to particle surface. PCL particles are used for controlled release, sustained release and targeted drug delivery. Degradation rate of PCL is slower than PLGA

Legal Information

Product of Phosphorex, Inc.Degradex(RM) is a product of Phosphorex, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric antioxidants show promise in drug delivery with superior bioavailability and antioxidant potential.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service