Skip to Content
Merck
All Photos(1)

Key Documents

805122

Sigma-Aldrich

PLGA microspheres

50 μm average diameter

Synonym(s):

Degradex® PLGA, PLGA Nanoparticles

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$658.00

$658.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$658.00

About This Item

Linear Formula:
(C3H4O2)x(C2H2O2)y
UNSPSC Code:
12162002
NACRES:
NA.23

$658.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

form

powder

Quality Level

average diameter

50 μm

density

1.3 g/cm3

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Degradex® is made with poly(D,L-lactide-co-glycolide) with an L/G ratio of 50/50 and a Mw of 30,000.
To re-constitute:
  • Remove the product from freezer.
  • Wait until temperature rises to the ambient temperature.
  • Add DI water or buffer to the container.
  • Vortex and sonicate for 5 minutes to ensure the microspheres are well suspended.

Application

Plain PLGA particles for testing system compatibility in drug delivery research. Particles can be used as carriers by attaching drug or biological/chemical entities to particle surface. PLGA particles are used for controlled release, sustained release and targeted drug delivery.

Legal Information

Degradex is a registered trademark of Phosphorex, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Study on how Degradex® PLGA microspheres and nanoparticles can be used for drug delivery research and formulation development.

Polymeric antioxidants show promise in drug delivery with superior bioavailability and antioxidant potential.

Biomaterials science integrates smart materials into biological research, requiring a deep understanding of biological systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service