Skip to Content
Merck
All Photos(1)

Key Documents

P8579

Sigma-Aldrich

1,3-Propanedisulfonic acid disodium salt

Synonym(s):

Eprodisate disodium, NC 503

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$248.00

$248.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$248.00

About This Item

Linear Formula:
NaSO3CH2CH2CH2SO3Na
CAS Number:
Molecular Weight:
248.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$248.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 07, 2025Details


Request a Bulk Order

form

powder

SMILES string

[Na].OS(=O)(=O)CCCS(O)(=O)=O

InChI

1S/C3H8O6S2.2Na/c4-10(5,6)2-1-3-11(7,8)9;;/h1-3H2,(H,4,5,6)(H,7,8,9);;/q;2*+1/p-2

InChI key

DKGJFKPIUSHDIT-UHFFFAOYSA-L

Application

Reactant involved in synthesis of:
  • Cadmium(II) bipyridine complex metal-organic frameworks with alkanedisulfonates
  • Alkylenedisulfonate inorganic-organic hybrid materials

Glycosaminoglycan mimetic involved in pharmacological studies for the treatment of amyloid A amyloidosis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service