Skip to Content
Merck
All Photos(3)

Documents

N605

Sigma-Aldrich

2,6-Naphthalenedisulfonic acid disodium salt

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(SO3Na)2
CAS Number:
Molecular Weight:
332.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

>300 °C (lit.)

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)c1ccc2cc(ccc2c1)S([O-])(=O)=O

InChI

1S/C10H8O6S2.2Na/c11-17(12,13)9-3-1-7-5-10(18(14,15)16)4-2-8(7)6-9;;/h1-6H,(H,11,12,13)(H,14,15,16);;/q;2*+1/p-2

InChI key

WZZLWPIYWZEJOX-UHFFFAOYSA-L

Other Notes

Remainder inorganic salts

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R M Wittich et al.
Applied and environmental microbiology, 54(7), 1842-1847 (1988-07-01)
A naphthalene-2,6-disulfonic acid (2,6NDS)-degrading Moraxella strain was isolated from an industrial sewage plant. This culture could also be adapted to naphthalene-1,6-disulfonic acid as growth substrate. Regioselective 1,2-dioxygenation effected desulfonation and catabolism to 5-sulfosalicylic acid (5SS), which also could be used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service