P3640
Poly(ethylene glycol)
average mol wt 3015-3685
Synonym(s):
PEG
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
Recommended Products
mol wt
average mol wt 3015-3685
SMILES string
C(CO)O
InChI
1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2
InChI key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Part of an aqueous two-phase system for the fractionation of proteins.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Anna E V Hagström et al.
PloS one, 10(5), e0126571-e0126571 (2015-05-16)
Noroviruses are recognized worldwide as the principal cause of acute, non-bacterial gastroenteritis, resulting in 19-21 million cases of disease every year in the United States. Noroviruses have a very low infectious dose, a short incubation period, high resistance to traditional
Akiko Harada et al.
Plants (Basel, Switzerland), 9(3) (2020-03-22)
In mesophyll cells of the aquatic monocot Vallisneria, red light induces rotational cytoplasmic streaming, which is regulated by the cytoplasmic concentration of Ca2+. Our previous investigations revealed that red light induces Ca2+ efflux across the plasma membrane (PM), and that
D J Morré et al.
BioTechniques, 7(9), 946-948 (1989-10-01)
Plasma membranes are readily purified from crude mixtures by the technique of aqueous two-phase partition. This procedure has been used widely to prepare plasma membrane fractions, highly purified, from both green and dark-grown plant materials. Only recently, however, has the
Larsson, C, et al.
Methods in Enzymology, 148, 558-568 (1987)
Colin J B Harvey et al.
Science advances, 4(4), eaar5459-eaar5459 (2018-04-14)
For decades, fungi have been a source of U.S. Food and Drug Administration-approved natural products such as penicillin, cyclosporine, and the statins. Recent breakthroughs in DNA sequencing suggest that millions of fungal species exist on Earth, with each genome encoding
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service