Skip to Content
Merck
All Photos(2)

Documents

P35359

Sigma-Aldrich

Diphenyl sulfone

97%

Synonym(s):

Phenyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2SO2
CAS Number:
Molecular Weight:
218.27
Beilstein:
1910573
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

379 °C (lit.)

mp

123-129 °C (lit.)

SMILES string

O=S(=O)(c1ccccc1)c2ccccc2

InChI

1S/C12H10O2S/c13-15(14,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H

InChI key

KZTYYGOKRVBIMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

363.2 °F

Flash Point(C)

184 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles W Locuson et al.
Journal of medicinal chemistry, 50(6), 1158-1165 (2007-02-22)
Several laboratories have demonstrated that activation of drug metabolism by P450s may occur via a mechanism that resembles allosterism from an enzyme kinetic standpoint. Because the effector drug binding site may be located in the same P450 binding pocket where
Anita Toulmin et al.
Journal of medicinal chemistry, 51(13), 3720-3730 (2008-06-19)
Partition coefficients were measured for 47 compounds in the hexadecane/water ( P hxd) and 1-octanol/water ( P oct) systems. Some types of hydrogen bond acceptor presented by these compounds to the partitioning systems are not well represented in the literature
Giulia Caron et al.
Journal of medicinal chemistry, 48(9), 3269-3279 (2005-04-29)
Growing interest in the use of both the logarithm of the partition coefficient of the neutral species in the alkane/water system (log P(N)(alk)) and the difference between log P(N)(oct) (the logarithm of the partition coefficient of the neutral species in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service