Merck
  • Use of simple docking methods to screen a virtual library for heteroactivators of cytochrome P450 2C9.
Use of simple docking methods to screen a virtual library for heteroactivators of cytochrome P450 2C9.
Journal of medicinal chemistry (2007-02-22)
Charles W Locuson, Peter M Gannett, Robyn Ayscue, Timothy S Tracy
ABSTRACT
Several laboratories have demonstrated that activation of drug metabolism by P450s may occur via a mechanism that resembles allosterism from an enzyme kinetic standpoint. Because the effector drug binding site may be located in the same P450 binding pocket where the drug substrate is located, the ability to find and characterize novel effectors (aka heteroactivators) will prove to be important in probing the mechanism of activation. We have used analogues of the prototypical CYP2C9 heteroactivator dapsone to validate a simple docking method that can be used to predict heteroactivators based on ligand binding location in a P450 crystal structure. As proof of concept for the described docking method, a protocol was developed to discover potential heteroactivators from a virtual chemical library through efficient sorting of >40,000 compounds. One of the top-scoring compounds identified was verified to be a CYP2C9 heteroactivator in vitro, and it possessed activity similar to dapsone.
Product Description
Supelco
Sulfamethoxazole, VETRANAL®, analytical standard
Supelco
Sulfadiazine, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfadiazine, 99.0-101.0%
Supelco
Sulfamethoxazole
Supelco
Dapsone, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)
Sigma-Aldrich
Sulfanilamide, ≥98%
Supelco
Dibenzothiophene, analytical standard
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Sigma-Aldrich
Dibenzothiophene, ≥99%
Sigma-Aldrich
Diphenyl sulfide, 98%
Sigma-Aldrich
4,4′-Sulfonyldiphenol, 98%
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Supelco
Diphenyl sulfone, PESTANAL®, analytical standard
Supelco
Sulfamethazine, VETRANAL®, analytical standard
Sigma-Aldrich
Dibenzothiophene, 98%