Skip to Content
Merck
All Photos(1)

Key Documents

232750

Sigma-Aldrich

1-Bromo-2-chloroethane

98%

Synonym(s):

Ethylene bromochloride, Ethylene chlorobromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$42.40
250 G
$179.00

$42.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$42.40
250 G
$179.00

About This Item

Linear Formula:
ClCH2CH2Br
CAS Number:
Molecular Weight:
143.41
Beilstein:
605265
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$42.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.488 (lit.)

bp

106-107 °C (lit.)

mp

−18-−14 °C (lit.)

solubility

water: soluble

density

1.723 g/mL at 25 °C (lit.)

functional group

bromo
chloro

SMILES string

ClCCBr

InChI

1S/C2H4BrCl/c3-1-2-4/h1-2H2

InChI key

IBYHHJPAARCAIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Dissociative double ionization and multi-photon ionization of 1-bromo-2-chloroethane (BCE) irradiated by the 800nm femtosecond laser field has been investigated by dc-slice imaging technology[1]. Conformational properties of BCE in several zeolites such as silicalite-1, siliceous Y, Na-Y and zeolite L have been investigated by FT-Raman spectroscopy[2].

Application

1-Bromo-2-chloroethane was used in regio- and diastereoselective synthesis of 2-alkylidenetetrahydrofurans[3].

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haiyan Wang et al.
Langmuir : the ACS journal of surfaces and colloids, 25(14), 8042-8050 (2009-04-21)
The conformational properties of 1-bromo-2-chloroethane (BCE) in several representative zeolites including silicalite-1, siliceous Y (Si-Y), Na-Y, and zeolite L have been investigated by FT-Raman spectroscopy in combination with thermogravimetric analysis. The results indicate that the conformational and dynamic behavior of
Yan Yang et al.
The Journal of chemical physics, 135(6), 064303-064303 (2011-08-17)
The dissociative double ionization and multi-photon ionization of 1-bromo-2-chloroethane (BCE) irradiated by the 800 nm femtosecond laser field have been investigated by dc-slice imaging technology. The charged parent ion ratio [BCE(2+)]/[BCE(+)] was measured, and the corresponding ionization process including non-sequential
P Langer et al.
The Journal of organic chemistry, 66(18), 6057-6063 (2001-09-01)
The domino C,O-cyclodialkylation reaction of dilithiated 1,3-dicarbonyl compounds with 1,4-dibromo-2-butene resulted in regio- and diastereoselective formation of 2-alkylidene-5-vinyltetrahydrofurans. The cyclization of 1,3-dicarbonyl dianions with 1-bromo-2-chloroethane regio- and diastereoselectively afforded 2-alkylidenetetrahydrofurans under thermodynamic reaction control.
L A Ballering et al.
Mutation research, 285(2), 209-217 (1993-02-01)
The genetic activity profiles of three structurally related dihaloalkanes, 1,2-dibromoethane (DBE), 1,2-dichloroethane (DCE) and 1-bromo-2-chloroethane (BCE), were compared in germ cells and somatic tissue of Drosophila melanogaster. The two genotoxicity indices estimated after mutagen exposure of male germ cells were
Irina Yankelzon et al.
Environmental science and pollution research international, 27(18), 22749-22757 (2020-04-24)
Multi-elemental C-Br-Cl compound-specific isotope analysis was applied for characterizing abiotic and biotic degradation of the environmental pollutant 1-bromo-2-chloroethane (BCE). Isotope effects were determined in the model processes following hydrolytic dehalogenation and dihaloelimination pathways as well as in a microcosm experiment

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service