Skip to Content
Merck
All Photos(3)

Key Documents

P2604

Sigma-Aldrich

Pentachlorophenol

97%

Synonym(s):

1-Hydroxy-2,3,4,5,6-pentachlorobenzene, 1-Hydroxypentachlorobenzene, 2,3,4,5,6-Pentachlorophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6Cl5OH
CAS Number:
Molecular Weight:
266.34
Beilstein:
1285380
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

9.2 (vs air)

Quality Level

vapor pressure

40 mmHg ( 211.2 °C)

Assay

97%

form

powder or chunks

bp

310 °C (lit.)

mp

165-180 °C (lit.)

density

1.978 g/mL at 25 °C (lit.)

SMILES string

Oc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl

InChI

1S/C6HCl5O/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H

InChI key

IZUPBVBPLAPZRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoonsang Cho et al.
Journal of medicinal chemistry, 51(19), 5984-5992 (2008-09-10)
HisG is an ATP-phosphoribosyl transferase (ATPPRTase) that catalyzes the first step in the biosynthetic pathway for histidine. Among the enzymes in this pathway, only HisG represents a potential drug target for tuberculosis. Only a few inhibitors with limited potency for
P G Jorens et al.
Human & experimental toxicology, 12(6), 479-495 (1993-11-01)
Pentachlorophenol (PCP) was, and still is, one of the most frequently used fungicides and pesticides. Its toxicity is due to interference with oxidative phosphorylation. Acute and chronic poisoning may occur by dermal absorption, inhalation or ingestion. Chronic poisoning occurs mainly
Glinda S Cooper et al.
Environmental health perspectives, 116(8), 1001-1008 (2008-08-19)
Pentachlorophenol, a fungicide widely used as a wood preservative, was classified in 1999 by the International Agency for Research on Cancer as a possible human carcinogen. We reviewed currently available data to determine the extent to which recent studies assist
M B Carvalho et al.
Journal of proteomics, 78, 159-171 (2012-11-28)
Pentachlorophenol (PCP) represents a critical concern worldwide due to its toxicity and recalcitrance to degradation. The capacity of Mucor plumbeus to transform PCP into several detoxification metabolites, including tetrachlorohydroquinone and several phase II conjugates, was observed by LC-HRMS. The data
Weiwei Zheng et al.
Environment international, 42, 105-116 (2011-05-24)
Pentachlorophenol (PCP) has been widely used for killing snails in areas of China where schistosomiasis is epidemic. With the re-emergence of schistosomiasis, the warranted production and consumption of PCP has inevitably resulted in persistent environmental contamination by it and its

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service