Key Documents
MT30130862
Mettler Toledo SevenCompact pH meter S210
Meter Only
Synonym(s):
ph meter, sevencompact ph meter
About This Item
UNSPSC Code:
41115603
NACRES:
NB.61
AC/DC input
12 V, universal plug set (mains adaptor)
description
connector BNC/RCA
interface RS232, USB-A, USA-B communication
pH/ion/ORP meters
feature
Meter Only
manufacturer/tradename
Mettler-Toledo 30130862
mV measuring range
-2000.0-2000.0
pH measuring range
-2.000-20.000
temperature measuring range
-30.0-130.0 °C (MTC)
-5.0-130.0 °C (ATC)
General description
This universal well designed meter is the perfect choice for a wide range of applications without overstretching your budget. It is much more than a pH meter: it can also measure ORP.
The included electrode holder with the perfectly vertical uPlace movement helps you bring your electrode in the position where it works best for your sample. This allows for faster measurements and reduces the risk of the sample vessel tipping over and/or damage the sensor.
SevenCompact gives you the choice to view all measurement information on the display or to focus on the essentials with the uFocus view. Switching between the different display layouts is easy, requiring only a single keypress.
The new color display with its well-designed icons and menu settings in 10 languages makes operation genuinely intuitive. Applications range from routine measurements through sample analysis, data handling and data archiving in compliance with GLP regulations. Simply innovative design that meets the requirements for a universal, easy-to-operate pH meter.
  • User friendly instrument for demanding users
  • Security and high reproducibility thanks to Intelligent Sensor Management (ISM®)
  • Improved quality of measurements through professional calibration support
  • Comprehensive service package including IQ/OQ
  • Integrated USB and RS232 interfaces for data exchange
Features and Benefits
Includes: Meter, electrode holder, protective cover, instructions, quick guide, test certificate, declaration of conformity
Legal Information
ISM is a registered trademark of Mettler-Toledo, Inc.
SevenCompact is a trademark of Mettler-Toledo, Inc.
uFocus is a trademark of Mettler-Toledo, Inc.
uPlace is a trademark of Mettler-Toledo, Inc.