MT30046245
Mettler-Toledo SevenExcellence™ conductivity meter S700
S700 standard kit, with InLab® 731-ISM, AC/DC input 230 V AC, universal plug set
About This Item
Recommended Products
description
Conductivity range 0.001 ?S/cm to 2000 ms/cm, various different units, defined by user (+/- 0.5%)
Resistivity range 0.01 ... 100.0 MOhm*cm
feature
with InLab® 731-ISM
manufacturer/tradename
Mettler-Toledo 30046245
salinity measuring range
0-80 ppt
temperature measuring range
-30-130 °C, accuracy: ±0.1
Looking for similar products? Visit Product Comparison Guide
General description
Professional, yet intuitive instrument with touchscreen operation, which can easily handle the most complex applications and meets stringent requirements in regulated markets.
- Flexible due to various conductivity modes, units and temperature correction possibilities
- Large 7 inch color display and intuitive menu guidance in 10 languages
- Flexible method concept for high reproducibility and security
- Extra security thanks to sophisticated user management and Intelligent Sensor Management (ISM®)
- Integrated interfaces (USB, RS232 and Ethernet) for data exchange
- Comprehensive service package including IQ/OQ/PQ
uPlace™ electrode holder with a perfect vertical motion. It can be operated with one hand and used either freestanding or attached to the left or right side of the instrument.
LabX direct pH PC software for transferring real-time measurement data and archiving your results
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service