Skip to Content
Merck
All Photos(1)

Key Documents

M80709

Sigma-Aldrich

Methyl stearate

97%

Synonym(s):

Methyl octadecanoate, Stearic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€380.40
250 MG
€527.30
500 MG
€802.90
1 G
€1,017.40

Vendor SKU: EN300-179218-100MG

€380.40


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
€380.40
250 MG
€527.30
500 MG
€802.90
1 G
€1,017.40

About This Item

Linear Formula:
CH3(CH2)16CO2CH3
CAS Number:
Molecular Weight:
298.50
Beilstein:
1786213
EC Number:
MDL number:
UNSPSC Code:
12352211

Vendor SKU: EN300-179218-100MG

€380.40


Ships in 14 days - from Aldrich Partner.

Assay

97%

bp

181-182 °C/4 mmHg (lit.)

mp

37-41 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C19H38O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h3-18H2,1-2H3

InChI key

HPEUJPJOZXNMSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Jian-Rong Wang et al.
    Journal of microbiology and biotechnology, 25(6), 845-855 (2015-01-16)
    The present study describes the gene cloning and high-level expression of an alkaline and thermostable lipase gene from Trichosporon coremiiforme V3. Nucleotide analysis revealed that this lipase gene has an open reading frame of 1,692 bp without any introns, encoding
    Sonja Klee et al.
    Rapid communications in mass spectrometry : RCM, 28(14), 1591-1600 (2014-06-05)
    Atmospheric pressure chemical ionization (APCI) sources operated with point to plane DC discharges ('Coronas') frequently suffer from point electrode degradation and potentially lead to oxidation and/or fragmentation of the generated analyte ions. It is postulated that these adverse effects are
    Preeti B Subhedar et al.
    Ultrasonics sonochemistry, 27, 530-535 (2015-05-03)
    The synthesis of biodiesel from sunflower oil and methanol based on transesterification using the immobilized lipase from Thermomyces lanuginosus (Lipozyme TL-IM) has been investigated under silent conditions and under an ultrasound field. Ultrasound assisted process led to reduced processing time
    Hynek Balcar et al.
    Beilstein journal of organic chemistry, 11, 2087-2096 (2015-12-15)
    Hoveyda-Grubbs type catalysts with cationic tags on NHC ligands were linker-free immobilized on the surface of lamellar zeolitic supports (MCM-22, MCM-56, MCM-36) and on mesoporous molecular sieves SBA-15. The activity of prepared hybrid catalysts was tested in olefin metathesis reactions:
    Obie Farobie et al.
    Bioresource technology, 191, 306-311 (2015-05-26)
    In this study, biodiesel production under supercritical conditions among methanol, ethanol, and tert-butyl methyl ether (MTBE) was compared in order to elucidate the differences in their reaction behavior. A continuous reactor was employed, and experiments were conducted at various reaction

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service