Skip to Content
Merck
All Photos(1)

Key Documents

M4805

Sigma-Aldrich

2-Mercapto-4-methylpyrimidine hydrochloride

99%

Synonym(s):

4-Methyl-2-pyrimidinethiol hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
CHF 628.30

Vendor SKU: MS-2169-500MG

CHF 628.30


Availability
Ships in 21 days - from Aldrich Partner.

Request a Bulk Order

Select a Size

Change View
500 MG
CHF 628.30

About This Item

Empirical Formula (Hill Notation):
C5H6N2S · HCl
CAS Number:
Molecular Weight:
162.64
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Vendor SKU: MS-2169-500MG

CHF 628.30


Availability
Ships in 21 days - from Aldrich Partner.

Request a Bulk Order

Assay

99%

mp

265 °C (dec.) (lit.)

SMILES string

Cl[H].Cc1ccnc(S)n1

InChI

1S/C5H6N2S.ClH/c1-4-2-3-6-5(8)7-4;/h2-3H,1H3,(H,6,7,8);1H

InChI key

UQJLPBLXSJWAKG-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service