Skip to Content
Merck
All Photos(1)

Key Documents

M38400

Sigma-Aldrich

2-Methylcyclohexanone

99%

Synonym(s):

Tetrahydro-o-cresol

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$90.30
100 ML
$109.00

$90.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$90.30
100 ML
$109.00

About This Item

Linear Formula:
CH3C6H9(=O)
CAS Number:
Molecular Weight:
112.17
Beilstein:
506751
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$90.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.448 (lit.)

bp

162-163 °C (lit.)

density

0.924 g/mL at 25 °C (lit.)

SMILES string

CC1CCCCC1=O

InChI

1S/C7H12O/c1-6-4-2-3-5-7(6)8/h6H,2-5H2,1H3

InChI key

LFSAPCRASZRSKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

118.4 °F

Flash Point(C)

48 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Halis T Balaydın et al.
Bioorganic & medicinal chemistry letters, 22(3), 1352-1357 (2012-01-11)
The Naturally occurring novel cyclohexanonyl bromophenol 2(R)-2-(2,3,6-tribromo-4,5-dihydroxybenzyl)cyclohexanone (4) was synthesized as a racemic compound. Cyclohexylphenyl methane derivatives (10-17) with Br, OMe, CO, and OH were also obtained. Inhibition of four human carbonic anhydrase (hCA, EC 4.2.1.1) isozymes I, II, IV

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service