Skip to Content
Merck
All Photos(2)

Key Documents

M34804

Sigma-Aldrich

Methyl 4-chlorobutyrate

≥98%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$94.40

$94.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$94.40

About This Item

Linear Formula:
ClCH2CH2CH2COOCH3
CAS Number:
Molecular Weight:
136.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$94.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98%

refractive index

n20/D 1.433 (lit.)

bp

175-176 °C (lit.)

density

1.12 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CCCCl

InChI

1S/C5H9ClO2/c1-8-5(7)3-2-4-6/h2-4H2,1H3

InChI key

ZZUYIRISBMWFMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service