Skip to Content
Merck
All Photos(1)

Key Documents

C29835

Sigma-Aldrich

4-Chlorobutyric acid

99%

Synonym(s):

4-Chlorobutanoic acid, 4-Chlorobutyrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$153.00
100 G
$373.00

$153.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$153.00
100 G
$373.00

About This Item

Linear Formula:
Cl(CH2)3COOH
CAS Number:
Molecular Weight:
122.55
Beilstein:
1700264
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$153.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.451 (lit.)

bp

196 °C/22 mmHg (lit.)

mp

12-16 °C (lit.)

density

1.24 g/mL at 25 °C (lit.)

SMILES string

OC(=O)CCCCl

InChI

1S/C4H7ClO2/c5-3-1-2-4(6)7/h1-3H2,(H,6,7)

InChI key

IPLKGJHGWCVSOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

<ul>
<li><strong>Application in hydrophilic fluorescent nanozymes: </strong>4-Chlorobutyric acid is used in the synthesis of Mn2+:CsPbCl3@SiO2 yolk shell nanoparticles (YSNPs) through the SN2 reaction with oleylamine (OAm). This reaction yields a zwitterionic ligand that facilitates the dispersion of YSNPs in water (Wang et al., 2024).</li>
</ul>

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Michele Dei Cas et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1154, 121982-121982 (2020-08-31)
Short and medium fatty acids derived from either dietary sources, gut microbiota, and liver production might play a role in the modulation of metabolism and inflammation. The outcome of different autoimmune or inflammatory diseases could be related to microbiota composition

Questions

  1. is it condutive or not ???? if is what is Coductivity????.

    1 answer
    1. The conductive properties of this material have not been investigated.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service