Skip to Content
Merck
All Photos(1)

Key Documents

A52005

Sigma-Aldrich

2-Amino-4,6-dimethylpyrimidine

95%

Synonym(s):

Acetylacetoneguanidine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$103.00

$103.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$103.00

About This Item

Empirical Formula (Hill Notation):
C6H9N3
CAS Number:
Molecular Weight:
123.16
Beilstein:
110735
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$103.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

95%

form

crystals

mp

151-153 °C (lit.)

SMILES string

Cc1cc(C)nc(N)n1

InChI

1S/C6H9N3/c1-4-3-5(2)9-6(7)8-4/h3H,1-2H3,(H2,7,8,9)

InChI key

IDQNBVFPZMCDDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christer B Aakeröy et al.
Pharmaceutics, 3(3), 601-614 (2011-01-01)
In the pharmaceutical industry, co-crystals are becoming increasingly valuable as crystalline solids that can offer altered/improved physical properties of an active pharmaceutical ingredient (API) without changing its chemical identity or biological activity. In order to identify new solid forms of
Yoshio Muguruma et al.
Biomedical chromatography : BMC, 34(2), e4722-e4722 (2019-10-28)
The metabolome presence of nucleobases, nucleosides, nucleotides and related phosphorylated metabolites has been examined for Alzheimer's disease (AD). Although reversed-phase liquid chromatography tandem mass spectrometry (LC-MS/MS) has been used for the determination of these analytes, they were limited in chromatographic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service