Skip to Content
Merck
All Photos(2)

Documents

340987

Sigma-Aldrich

4-(Trifluoromethoxy)benzylamine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4CH2NH2
CAS Number:
Molecular Weight:
191.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.452 (lit.)

bp

57-60 °C/10 mmHg (lit.)

density

1.252 g/mL at 25 °C (lit.)

SMILES string

NCc1ccc(OC(F)(F)F)cc1

InChI

1S/C8H8F3NO/c9-8(10,11)13-7-3-1-6(5-12)2-4-7/h1-4H,5,12H2

InChI key

DBGROTRFYBSUTR-UHFFFAOYSA-N

Related Categories

Application

4-(Trifluoromethoxy)benzylamine may be used in chemical synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon H Rüdisser et al.
Journal of biomolecular NMR, 74(10-11), 579-594 (2020-06-20)
Fluorine NMR has recently gained high popularity in drug discovery as it allows efficient and sensitive screening of large numbers of ligands. However, the positive hits found in screening must subsequently be ranked according to their affinity in order to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service