Skip to Content
Merck
All Photos(1)

Documents

L512206

Sigma-Aldrich

Ir(MDQ)2(acac)

AldrichCPR

Synonym(s):

Bis(2-methyldibenzo[f,h]quinoxaline)(acetylacetonate)iridium(III)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H29N4O2Ir
CAS Number:
Molecular Weight:
777.89
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

CC1=NC(C(C=CC=C2)=C2C3=C4C5=CC=C3)=C4[N]([Ir]56OC(C)C=C(C)O6)=C1

InChI

1S/C17H11N2.C5H9O2.Ir/c1-11-10-18-16-14-8-4-2-6-12(14)13-7-3-5-9-15(13)17(16)19-11;1-4(6)3-5(2)7;/h2-7,9-10H,1H3;3-4,7H,1-2H3;/q;-1;+2/p-1/b;5-3-;

InChI key

FQHZITKUOOFGDL-FMFSYIAASA-M

Looking for similar products? Visit Product Comparison Guide

General description

The product is an iridium-based electrophosphorescent emitter.

Application

Ir(MDQ)2(acac) exhibits orange red-electroluminescent transmission. Ir(MDQ)2(acac) doped in α - N,N-dinaphthalen-1-yl- N,N-diphenyl-benzidine (NPD) can act as host material in OLEDs.

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid intersystem crossing in highly phosphorescent iridium complexes
Tang KC, et al.
Chemical Physics Letters null
Influence of charge balance and exciton distribution on efficiency and lifetime of phosphorescent organic light-emitting devices.
Meerheim R, et al.
Journal of Applied Physics null
Quantification of energy loss mechanisms in organic light-emitting diodes.
Meerheim R, et al.
Applied Physics Letters, 253305 null
Rapid intersystem crossing in highly phosphorescent iridium complexes
Tang KC, et al.
Chemical Physics Letters, 437-441 null
Influence of charge balance and exciton distribution on efficiency and lifetime of phosphorescent organic light-emitting devices.
Meerheim R, et al.
Journal of Applied Physics, 014510 null

Articles

Professor Shinar highlights low-cost, disposable sensor configurations in organic and hybrid electronics for healthcare applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service