Skip to Content
Merck
All Photos(1)

Key Documents

JKA5146

Sigma-Aldrich

HS-PEG-NH2

average Mn 7,500, HCl Salt, cross-linking reagent thiol reactive, cross-linking reagent carboxyl reactive

Synonym(s):

HS-PEG-NH2, Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$640.00

$640.00


Estimated to ship onMay 08, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$640.00

About This Item

Linear Formula:
HS(CH2CH2O)nCH2CH2NH2HCl
UNSPSC Code:
12162002
NACRES:
NA.23

$640.00


Estimated to ship onMay 08, 2025


Request a Bulk Order

Product Name

HS-PEG7500-NH2, HCl Salt, average Mn 7,500

form

solid

mol wt

average Mn 7,500

reaction suitability

reagent type: cross-linking reagent
reactivity: carboxyl reactive

reagent type: cross-linking reagent
reactivity: thiol reactive

polymer architecture

shape: linear
functionality: heterobifunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Thiol PEG Amine. HS is thiol reactive and NH2 is stable and can be activated.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhang, Z., et al.
Biosensors And Bioelectronics, 59 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service