Skip to Content
Merck
All Photos(1)

Documents

J100005

Sigma-Aldrich

Bis(trimethylsilyl)amidohafnium(IV) chloride

≥95%, ≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[[(CH3)3Si]2N]2HfCl2
CAS Number:
Molecular Weight:
570.17
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

≥95%
≥99.99% trace metals basis

form

low-melting solid

reaction suitability

core: hafnium

mp

40-44 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)N([Si](C)(C)C)[Hf](Cl)(Cl)N([Si](C)(C)C)[Si](C)(C)C

InChI

1S/2C6H18NSi2.2ClH.Hf/c2*1-8(2,3)7-9(4,5)6;;;/h2*1-6H3;2*1H;/q2*-1;;;+4/p-2

InChI key

HKZRZVQUSXQGFM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 72 Hafnium

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Thin film photovoltaic devices have become increasingly important in efficiently harnessing solar energy to meet consumer demand.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service