Skip to Content
Merck
All Photos(1)

Key Documents

923451

Sigma-Aldrich

(PhMPhos)PdCl2

complex with CH2Cl2

Synonym(s):

1-Diadamantylphosphino-1′-diphenylphosphinoferrocene palladium dichloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$107.00
500 MG
$321.00

$107.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025



Select a Size

Change View
100 MG
$107.00
500 MG
$321.00

About This Item

Empirical Formula (Hill Notation):
C42H48Cl2FeP2Pd
CAS Number:
Molecular Weight:
847.95
UNSPSC Code:
12352101
NACRES:
NA.21

$107.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Quality Level

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

SMILES string

ClCCl.[Cl-][Pd+2]1([Cl-])[P](C=2C=CC=CC2)(C=3C=CC=CC3)[C-]45[CH]6=[CH]7[CH]8=[CH]4[Fe+2]789%10%11%1265[CH]=%13[CH]%12=[CH]%11[C-]%10([CH]%139)[P]1(C%14%15CC%16CC(CC(C%16)C%14)C%15)C%17%18CC%19CC(CC(C%19)C%17)C%18

InChI

InChI=1S/C25H34P.C17H14P.CH2Cl2.2ClH.Fe.Pd/c1-2-4-23(3-1)26(24-11-17-5-18(12-24)7-19(6-17)13-24)25-14-20-8-21(15-25)10-22(9-20)16-25;1-3-9-15(10-4-1)18(17-13-7-8-14-17)16-11-5-2-6-12-16;2-1-3;;;;/h1-4,17-22H,5-16H2;1-14H;1H2;2*1H;;/q2*-1;;;;2*+2/p-2

InChI key

CPZZSCIFYJFGRL-UHFFFAOYSA-L

Application

(PhMPhos)PdCl2 is an unsymmetrical ferrocene-based Pd-catalyst. The MPhos series of ligands are unsymmetrical diphosphino-ferrocenes bearing a bulky adamantyl group. (PhMPhos)PdCl2 can be used for a variety of cross-coupling transformations, including Suzuki coupling, C-P coupling, and Csp2-Csp3 coupling. Demonstrated Csp2-Csp3 couplings include Murahashi-Feringa Coupling (Alkyl-Li), Kumada-Corriu coupling (Alkyl-MgX), Negishi coupling (Alkyl-ZnX) and Suzuki–Miyaura coupling (Alkyl-B(OR2)).

Learn more about MPhos ligands and catalysts

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service