Skip to Content
Merck
All Photos(2)

Key Documents

GF97410708

Carbon

foil, 250x250mm, thickness 1.0mm, flexible graphite, 99.8%

Synonym(s):

Activated charcoal, C 000330, Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
12141908
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥99.8%

form

foil

autoignition temp.

842 °F

manufacturer/tradename

Goodfellow 974-107-08

resistivity

1375 μΩ-cm, 20°C (graphite)

size × thickness

250 × 250 mm × 1.0 mm

mp

3550 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M C Ledbetter et al.
Proceedings of the National Academy of Sciences of the United States of America, 84(1), 85-88 (1987-01-01)
Cluster ions containing 50, 100, or 200 water molecules and one proton are accelerated to over 275 kV and impacted on thin carbon films on which tobacco mosaic virus has been dispersed. After bombardment the films are examined with a
Z oscillation of mean charges of energetic ions emerging from a carbon foil: Correlation with the shell structure of ions.
Shima et al.
Physical review. A, General physics, 39(8), 4316-4318 (1989-04-15)
Birtcher et al.
Physical review letters, 85(23), 4968-4971 (2000-12-02)
In situ transmission electron microscopy has been used to observe sputtered Au during Xe ion irradiation in transmission geometry. The sputtered Au was collected on an electron transparent carbon foil. Nanoparticles were observed on the collector foil after they were
Formation of Rydberg states in fast ions penetrating thin carbon-foil and gas targets.
Schiwietz et al.
Physical review letters, 59(14), 1561-1564 (1987-10-05)
Systematics of equilibrium charge distributions of ions passing through a carbon foil over the ranges Z=4-92 and E=0.02-6 MeV/u.
Shima et al.
Physical review. A, General physics, 40(7), 3557-3570 (1989-10-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service