Product Description
Carbon, foil, 300x300mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 150x150mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 250x250mm, thickness 1.0mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.25mm, rigid graphite, fine grain size, 99.997%
Carbon, foil, 25x25mm, thickness 0.25mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 150x150mm, thickness 1.0mm, flexible graphite, 99.8%
Carbon, foil, 6mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 25x25mm, thickness 0.15mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 100x100mm, thickness 5.0mm, rigid graphite, medium grain size, 99.5%
Carbon, foil, 5x10mm, thickness 2.0mm, hOpg
Carbon, foil, 150x150mm, thickness 1.0mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 8mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.2mm, pyrolytic graphite, 99.99%
Carbon, foil, 10x10mm, thickness 2.0mm, hOpg
Carbon, foil, 50x50mm, thickness 5.0mm, rigid graphite, medium grain size, 99.5%
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Carbon, foil, not light tested, 100x100mm, thickness 0.075mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.35mm, flexible graphite, 99.8%
Carbon, foil, 4mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.25mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, not light tested, 150x150mm, thickness 0.075mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon, foil, 12mm disks, thickness 0.5mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, not light tested, 150x150mm, thickness 0.125mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.5mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 10mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 12mm disks, thickness 1.0mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, not light tested, 300x300mm, thickness 0.075mm, flexible graphite, 99.8%