Skip to Content
Merck
All Photos(2)

Key Documents

GF60323442

Titanium

foil, thickness 1.0 mm, diameter 4 mm, high purity 99.999%

Synonym(s):

TI000433

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ti
CAS Number:
Molecular Weight:
47.87
MDL number:
UNSPSC Code:
12141746
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.999%

form

foil

autoignition temp.

860 °F

manufacturer/tradename

Goodfellow 603-234-42

resistivity

42.0 μΩ-cm, 20°C

diam.

4 mm

thickness

1.0 mm

bp

3287 °C (lit.)

mp

1660 °C (lit.)

density

4.5 g/mL at 25 °C (lit.)

SMILES string

[Ti]

InChI

1S/Ti

InChI key

RTAQQCXQSZGOHL-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF29996892GF40430719GF53839974
assay

99.999%

assay

99.999%

assay

99.999%

assay

99.999%

form

foil

form

foil

form

foil

form

foil

mp

1660 °C (lit.)

mp

1660 °C (lit.)

mp

1660 °C (lit.)

mp

1660 °C (lit.)

resistivity

42.0 μΩ-cm, 20°C

resistivity

42.0 μΩ-cm, 20°C

resistivity

42.0 μΩ-cm, 20°C

resistivity

42.0 μΩ-cm, 20°C

bp

3287 °C (lit.)

bp

3287 °C (lit.)

bp

3287 °C (lit.)

bp

3287 °C (lit.)

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service