Skip to Content
Merck
All Photos(1)

Key Documents

DWK63A53

Kimble® Disposable Pasteur Pipet

L 5.75 in., Borosilicate glass/PTFE

Synonym(s):

63A53

Sign Into View Organizational & Contract Pricing

Select a Size

1000 EACH
$151.00

$151.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1000 EACH
$151.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.04

$151.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

Borosilicate glass/PTFE

packaging

pkg of 1000 ea

manufacturer/tradename

Kimble® 63A53

L

5.75 in.

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

These Pasteur-type pipets are intended for one-time use in the transfer of small volumes.

Application

Bacteriology, immunology, hematology, and serology

Features and Benefits

Removes risk of cross-contamination of high value specimens and laboratory reagents.

Legal Information

KIMBLE is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service