Skip to Content
Merck
All Photos(1)

Key Documents

DWK374326

Wheaton® Polypropylene Nozzle & Glass Sinker

Synonym(s):

374326

Sign Into View Organizational & Contract Pricing

Select a Size

6 EACH
$62.50

$62.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
6 EACH
$62.50

About This Item

UNSPSC Code:
41104812
NACRES:
NB.43

$62.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

polypropylene nozzle (6 mm)

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 6 ea

manufacturer/tradename

WHEATON® 374326

Looking for similar products? Visit Product Comparison Guide

Application

Research, Development and Quality Control, Pharmaceutical and Biotechnology, Food and Beverage, Waste Water and Sample Collection

Features and Benefits

  • The nozzle connects to the pump tubing for dispensing of liquids
  • The glass sinker is used to secure tubing when drawing from a container

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service