Key Documents
DWK2515415-B
Kimble® Closed Top Linerless Polypropylene Screw Thread Cap
15-415, black screw cap
Select a Size
1 PKG
$1,000.00
$1,000.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 PKG
$1,000.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.17
$1,000.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
black screw cap
red polypropylene screw cap (Push fit)
packaging
pkg of 1 packs
manufacturer/tradename
Kimble® 2515415-B
fitting
thread
Application
Closure solution for vials and bottles
Features and Benefits
  • Economical, one-piece construction
  • Unique design provides exceptional sealing properties
  • Autoclavable
  • Designed from ASTM Specification E982
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)