Documents
DWK11-5200-300
Wheaton® Microliter Vials With Inserts Installed
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
clear vial
feature
vial type with Marking Patch with 11-0000-300 inserts
packaging
pkg of 1 packs
manufacturer/tradename
WHEATON® 11-5200-300
Application
Chromatography
Features and Benefits
  • Double ring finish for added closure performance.
  • Approved for LC & GC.
  • Where noted, Certified Class 10,000 clean room packed enhances data integrity.
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.