Skip to Content
Merck
All Photos(1)

Key Documents

D44952

Sigma-Aldrich

Dibutylamine

≥98%

Synonym(s):

N-butylbutan-1-amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)2NH
CAS Number:
Molecular Weight:
129.24
Beilstein:
506001
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
bp:
159 °C (lit.)
vapor pressure:
1.9 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

4.46 (vs air)

Quality Level

vapor pressure

1.9 mmHg ( 20 °C)

Assay

≥98%

form

liquid

autoignition temp.

594 °F

expl. lim.

10 %

refractive index

n20/D 1.417 (lit.)

pH

11.1 (20 °C, 1 g/L)

bp

159 °C (lit.)

mp

−62 °C (lit.)

density

0.767 g/mL at 25 °C (lit.)

SMILES string

CCCCNCCCC

InChI

1S/C8H19N/c1-3-5-7-9-8-6-4-2/h9H,3-8H2,1-2H3

InChI key

JQVDAXLFBXTEQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

  • Dibutylamine is extensively used in palladium-catalyzed cross-coupling with aryl halides to synthesize arylamines, popularly known as Buchwald–Hartwig amination.[1][2]
  • It can be used in the oxone-mediated annulation of 2-aminobenzamides and 1,2-diaminobenzenes to synthesize 2,3-dihydroquinazolin-4(1H)-ones and 1H-benzimidazoles, respectively.[3]
  • It can also be used in the one-pot multicomponent reactions to synthesize tetra- and penta-substituted polyfunctional dihydropyrroles.[4]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.9 °F - closed cup

Flash Point(C)

40.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxone-mediated annulation of 2-aminobenzamides and 1, 2-diaminobenzenes with sec-amines via imine-N-oxides: new syntheses of 2, 3-dihydroquinazolin-4 (1 H)-ones and 1 H-benzimidazoles.
Sriramoju V, et al.
New. J. Chem., 42(5), 3188-3191 (2018)
Simple, efficient catalyst system for the palladium-catalyzed amination of aryl chlorides, bromides, and triflates.
Wolfe J P, et al.
The Journal of Organic Chemistry, 65(4), 1158-1174 (2000)
Concise and versatile multicomponent synthesis of multisubstituted polyfunctional dihydropyrroles.
Zhu Q, et al.
Journal of Combinatorial Chemistry, 11(4), 685-696 (2009)
Aqueous hydroxide as a base for palladium-catalyzed amination of aryl chlorides and bromides.
Kuwano R, et al.
The Journal of Organic Chemistry, 67(18), 6479-6486 (2002)
Baiyi Xue et al.
Journal of mass spectrometry : JMS, 46(7), 689-695 (2011-07-12)
Characterization of glycosaminoglycans poses a challenge for current analytical techniques, as they are highly acidic, polydisperse and heterogeneous compounds. The purpose of this study is the separation and analysis of a partially depolymerized heparin-like glycosaminoglycan by on-line ion-pairing reversed-phase high-performance

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service