Skip to Content
Merck
All Photos(1)

Key Documents

03205

Sigma-Aldrich

N-Ethylbutylamine

≥98.0%

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$55.00

$55.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$55.00

About This Item

Linear Formula:
CH3CH2CH2CH2NHCH2CH3
CAS Number:
Molecular Weight:
101.19
Beilstein:
1731324
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$55.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

3.5 (vs air)

vapor pressure

18 mmHg ( 20 °C)

Assay

≥98.0%

refractive index

n20/D 1.405 (lit.)

bp

108 °C (lit.)

density

0.74 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCCCNCC
CCCCNCC

InChI

1S/C6H15N/c1-3-5-6-7-4-2/h7H,3-6H2,1-2H3

InChI key

QHCCDDQKNUYGNC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service