Skip to Content
Merck
All Photos(2)

Key Documents

143707

Sigma-Aldrich

trans-2,3-Dibromo-2-butene-1,4-diol

97%

Synonym(s):

2,3-Dibromo-2-butene-1,4-diol

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$106.00

$106.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$106.00

About This Item

Linear Formula:
HOCH2C(Br)=C(Br)CH2OH
CAS Number:
Molecular Weight:
245.90
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$106.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

112-114 °C (lit.)

SMILES string

OC\C(Br)=C(/Br)CO

InChI

1S/C4H6Br2O2/c5-3(1-7)4(6)2-8/h7-8H,1-2H2/b4-3+

InChI key

MELXIJRBKWTTJH-ONEGZZNKSA-N

Application


  • Synthesis and Characterization of Oligomer Bis (trans-2, 3-dibromo-4-hydroxy-2-butenyl) terephthalate as a Green Corrosion Inhibitor on Mild Steel in 1 MH 3 PO 4: This study discusses the conversion of PET waste into an oligomer using trans-2,3-dibromo-2-butene-1,4-diol, exploring its efficacy as a corrosion inhibitor (Tuama, Indonesian Journal of Chemistry).

  • Use of bromine and bromo-organic compounds in organic synthesis: This review highlights the role of trans-2,3-dibromo-2-butene-1,4-diol in organic synthesis, particularly its utility in bromination reactions to synthesize complex organic molecules (Saikia et al., Chemical Reviews, 2016).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service