Skip to Content
Merck
All Photos(1)

Key Documents

D186201

Sigma-Aldrich

2,3-Dimethylsuccinic acid

≥99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$125.00

$125.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$125.00

About This Item

Linear Formula:
HOOCCH(CH3)CH(CH3)COOH
CAS Number:
Molecular Weight:
146.14
Beilstein:
1723932
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$125.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥99%

form

powder

SMILES string

CC(C(C)C(O)=O)C(O)=O

InChI

1S/C6H10O4/c1-3(5(7)8)4(2)6(9)10/h3-4H,1-2H3,(H,7,8)(H,9,10)

InChI key

KLZYRCVPDWTZLH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Branlant
European journal of biochemistry, 121(2), 407-411 (1982-01-01)
Aldehyde reductase I from pig liver is strongly inhibited by cyclized NADP--2-oxodiacid adducts. This result, in conjunction with those showing a strong inhibitory effect of certain diacid derivatives, such as (+/-)-dimethylsuccinic acid, towards aldehyde reductase I, suggests the presence of
P S Thomas et al.
Postgraduate medical journal, 67(783), 63-65 (1991-01-01)
Chronic lead poisoning has traditionally been treated by parenteral agents. We present a case where a comparison of ethylene diaminetetra-acetic acid was made with 2,3-dimethyl succinic acid (DMSA) which has the advantage of oral administration associated with little toxicity and
E Asante-Appiah et al.
Biochemistry, 36(29), 8710-8715 (1997-07-22)
gem-Dimethylsuccinic acid and its higher homolog, 2-methyl-2-ethylsuccinic acid (MESA) are highly potent inhibitors of both carboxypeptidase A (CPA) and B. The inhibition constant of MESA for CPA (0.11 microM for the racemic mixture) is remarkable considering the relatively simple structure
P H Andersen et al.
Food additives and contaminants, 1(3), 283-288 (1984-07-01)
Three flavourings: dimethyl succinate, ethyl pyruvate and aconitic acid, commonly used in candy, beverages, and baked goods, were tested in the Salmonella/mammalian-microsome test. Tester strains were TA 1535, TA 100, TA 1537 and TA 98 and doses were 32, 160
Jannick Jacobsen et al.
Dalton transactions (Cambridge, England : 2003), 48(23), 8433-8441 (2019-05-23)
Six different chiral and achiral alkane dicarboxylic C4-acids, i.e. succinic acid (H2SUC), dl-2-methylsuccinic acid (H2MS), 2,3-dimethylsuccinic acid (H2DMS) and aspartic acid (d-, l- and dl-H2ASP), were used to obtain Ce(iv)-MOFs via microwave assisted reactions. In water-based syntheses, MOFs with three

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service